Learn More
Cole-Parmer™ PVC Check Valve, NPT(F)
Cole-Parmer™ PVC Check Valve, NPT(F) can be easily and quickly installed either vertically or horizontally.
Brand:  Cole-Parmer™ 01341-00
Additional Details : Weight : 0.80000kg
Product Code. 11882202
  • 585.00 DKK / Each
Estimated Shipment: 15-05-2024
to see stock.
Add to basket
Description
Description
  • Assembled with water soluble, silicone-free lubricant
  • Constructed of a one-piece PVC body
Specifications
Specifications
Check Valve
0.5 in. NPT Tubing
1 Pack
PVC
Chemical processing or water/wastewater applications
60°C
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products