Få mere at vide
Tocris Bioscience™ Purvalanol A
Artikelnummer. 18015652
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18015652 10 mg
18084372 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18015652 Leverandør Tocris Bioscience™ Leverandørnr. 1580/10
Cdk inhibitor; potently inhibits cdk1, cdk2 and cdk5
Cyclin-dependent kinase (cdk) inhibitor (reported IC50 values are 4, 4 - 70, 75 - 240 and 100 nM for cdk1, cdk2, cdk5 and cdk 7, respectively). Reversibly arrests synchronised cells in G1 and G2, and inhibits cell proliferation and cell death.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 212844-53-6
Molekylær formel C19H25ClN6O
Tekniske data
Kemisk navn eller materiale Purvalanol A
Mængde 10 mg
Formel vægt 388.9
Renhed >98%
Molekylvægt (g/mol) 388.90