Beskrivelse
- Made with a rigid polycarbonate frame enclosing polypropylene reaction wells
- Available in styles to fit major cyclers and are freezable (-80°C)
- Choose from 96 or 384 well capacity
Tekniske data
Tekniske data
| Stregkode | No Barcode |
| Certificeringer/compliance | ISO 9001:2015, FDA CFR 21 GMP |
| Format | SBS Standard |
| Array | 12 x 8 |
| Autoklaverbar | Not Autoclavable |
| Emballage | 10 Plates/Pk., 10 Pk./Cs. |
| Farve | Clear |
| Engangs | Single-use |
| Produkttype | PCR plate |
| Nederdel stil | Semi skirt |
| Vis mere |