Få mere at vide
Beskrivelse
The pump bottle with dispenser delivers an accurate, clean and dosed amount of the medium. It is excellent for a variety of liquids such as reagents, detergents, disinfectants or lotions.The dispenser pump protects the contents from contamination and allows hygienic dispensing as the pump minimises air and germ ingress and prevents fingers or other objects from coming into contact with the contents. The crystal clear empty bottle fits comfortably in the hand and allows the contents to be clearly seen. It can be used multiple times and provides efficient residue emptying. The pump head of the dispenser pump can be locked in place with a twist-and-lock cap, ensuring that no liquid can escape.
- Dispenses a precise amount
- Protects the contents
- Tightly closes with extra long closure thread
Tekniske data
Tekniske data
| Kapacitet (metrisk) | 100 mL |
| Kapacitet (engelsk) | 3.38 fl oz. |
| Farve | White |
| Materiale | PET |
| Sterilitet | Non-sterile |
| Lukning inkluderet | Yes |
| Engangs | Reusable |
| Uddannet | No |
| Mund | Narrow |
| Form | High Shoulder |
| Vis mere |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.