missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PUMA Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0128
Additional Details : Weight : 0.01000kg
Description
PUMA Synthetic Peptide, PEP-0128, from Invitrogen.
PUMA is one of the pro-apoptotic Bcl-2 family members including Bax and Noxa, which are also transcriptional targets of p53. The PUMA gene encodes two BH3 domain-containing proteins termed PUMA-a and PUMA-b. PUMA proteins bind Bcl-2, localize to the mitochondria, and induce cytochrome c release and apoptosis in response to p53. PUMA may be a direct mediator of p53-induced apoptosis. Antisense inhibition of PUMA expression reduced the apoptotic response to p53, and PUMA is likely to play a role in mediating p53-induced cell death through the cytochrome c/Apaf-1-dependent pathway. This peptide corresponds to 15 amino amino terminus of human PUMA.PEP-0128 can be used as a blocking peptide with polyclonal antibody PA5-20008.Specifications
27113 | |
purified | |
Human PUMA | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q96PG8 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
BBC3 | |
0.02% sodium azide | |
50 μg | |
PBS |