missing translation for 'onlineSavingsMsg'
Få mere at vide

PTPRN2 Antibody [Alexa Fluor« 647], Novus Biologicals Biologicals™

Artikelnummer. 30498939 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.1 mL
Pakningsstørrelse:
0.1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30498939 0.1 mL 0.1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30498939 Supplier Novus Biologicals Supplier No. NBP335468AF647

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PTPRN2 Polyclonal antibody specifically detects PTPRN2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTPRN2
Applikationer ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugeret Alexa Fluor 647
Formulering 50mM Sodium Borate
Gene Alias EC 3.1.3.48, IA-2beta, IAR/receptor-like protein-tyrosine phosphatase, ICAARIARPTPRP, Islet cell autoantigen-related protein, KIAA0387, phogrin, protein tyrosine phosphatase receptor pi, protein tyrosine phosphatase, receptor type, N polypeptide 2, receptor-type tyrosine-protein phosphatase N2, R-PTP-N2, tyrosine phosphatase IA-2 beta
Værtsarter Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 22-242 of human PTPRN2 (NP_570858.2).,, Sequence:, AAPSSVPRGRQLPGRLGCLLEEGLCGASEACVNDGVFGRCQKVPAMDFYRYEVSPVALQRLRVALQKLSGTGFTWQDDYTQYVMDQELADLPKTYLRRPEASSPARPSKHSVGSERRYSREGGAALANALRRHLPFLEALSQAPASDVLARTHTAQDRPPAEGDDRFSESILTYVAHTSALTYPPGSRTQLREDLLPRTLGQLQPDELSPKVDSGVDRHHL
Oprensningsmetode Affinity purified
Mængde 0.1 mL
Regulatorisk status RUO
Forskningsdisciplin Lipid and Metabolism, Protein Phosphatase
Primær eller sekundær Primary
Gen-id (Entrez) 5799
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4°C in the dark.
Produkttype Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.