Få mere at vide
Novus Biologicals™ PTPN20 Recombinant Protein Antigen
Artikelnummer. 18130719 Shop alle Bio Techne produkter
Skift visning
Protein længde:
MALELKNLPGEFNSGNQPSNREKNRYRDILP
NFPHNDYEDVFEEPSESGSDPSMWTARG
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Protein længde
18130719 NFPHNDYEDVFEEPSESGSDPSMWTARG
18134779 MALELKNLPGEFNSGNQPSNREKNRYRDILP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18130719 Leverandør Novus Biologicals™ Leverandørnr. NBP246658PEP
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human PTPN20 The PTPN20 Recombinant Protein Antigen is derived from E. coli. The PTPN20 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
TRUSTED_SUSTAINABILITY
Tekniske data
Protein længde NFPHNDYEDVFEEPSESGSDPSMWTARG
Renhed >80% by SDS-PAGE and Coomassie blue staining
Fælles navn PTPN20
Indhold og opbevaring Store at −20°C short term. Avoid freeze-thaw cycles.
Formulering PBS and 1M Urea, pH 7.4
Til brug med (applikation) AC
Gene Alias protein tyrosine phosphatase, non-receptor type 20, bA42B19.1, protein tyrosine phosphatase, non-receptor type 20B
Gen symbol PTPN20
Etikettype Unlabeled
Molekylvægt (g/mol) 21 kDa
Produkttype Recombinant Protein Antigen
Mængde 0.1 mL
Regulatorisk status RUO
Kilde E.coli
Specifik reaktivitet Human
Vis mere Vis mindre
For Research Use Only.