missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PTK2B (FAK2) Recombinant Human Protein
PTK2B (FAK2) is a cytoplasmic tyrosine kinase.
4375.00 DKK - 57500.00 DKK
Specifications
Accession Number | NP_004094 |
---|---|
Formulation | 50mM Tris (pH 7.5), 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 145.1kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10352213
|
Invitrogen™
PV4567 |
10μg |
4375.00 DKK
10µg |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15498653
|
Invitrogen™
PR8161B |
100μg |
13290.00 DKK
100µg |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15488733
|
Invitrogen™
PV4568 |
1mg |
57500.00 DKK
1mg |
Estimated Shipment: 18-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PTK2B (FAK2) is involved in calcium-induced regulation of ion channels and activation of the map kinase signaling pathway.
Specifications
NP_004094 | |
145.1kDa | |
RUO - research use only | |
FAK Kinase Family | |
Recombinant | |
Baculovirus |
50mM Tris (pH 7.5), 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
PTK2B (FAK2) | |
Human | |
GST Tag |