Få mere at vide
SVL™ Rotulex™ PTFE O-ring til kugleled
Borestørrelse:
5 mm
9 mm
15 mm
20 mm
25 mm
40 mm
Pakningsstørrelse:
Hver
Description
- Offers a grease-free spherical joint
- PTFE pathway is highly resistant to chemical corrosion
- Can be used at temperatures up to 200°C
- PTFE covered O-ring fits into a female cup joint
- Joint can be retained together with spring loaded metal clip
Specifications
Specifications
| Borestørrelse | 40 mm |
| Til brug med (udstyr) | RM:RF64 Glass Joint |
| Længde (metrisk) | 45 mm Shank |
| Mængde | 1 Ea. |
| Diameter (metrisk) Udvendig | 64 mm |
| Materiale | PTFE |
| Type | Seal Ring |