Learn More
Fisherbrand™ Octagonal Magnetic Stir Bar Assortment with Retainer
Provides greater surface area and added turbulence when compared to smooth cylindrical bars. Fisherbrand™ Octagonal Magnetic Stir Bar Assortments with Restrainers have an integral pivot ring on each bar to aid in reducing friction and chattering.
Brand: Fisherbrand™
482.56 DKK valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 0.80000kg
Description
- Alnico magnets encapsulated in FDA grade low friction coating
- Restrainer fits comfortably in one hand
- Eliminates splashing hazards when pouring
Order Info
DimensionsLxW
Specifications
Integral Pivot Rings, Spinbar Restrainer | |
12.7 x 3.175, 12.7 x 7.92, 38.1 x 7.92, 50.8 x 7.92mm | |
1/Ea. | |
ISO 9001:2008 |
PTFE | |
Magnetic Stir Bar, Micro | |
PTFE | |
Octagon |