Få mere at vide
Pterostilbene, TRC
Artikelnummer. 30012908
Skift visning
Mængde:
50 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012908 1 g
30009646 50 mg
30011975 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012908 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8398901G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 537-42-8
Molekylær formel C16 H16 O3
Molekylvægt (g/mol) 256.3
Synonym Phenol, 4-[(1E)-2-(3,5-dimethoxyphenyl)ethenyl]-, 4-Stilbenol, 3',5'-dimethoxy-, (E)- (8CI), Phenol, 4-[2-(3,5-dimethoxyphenyl)ethenyl]-, (E)-, (E)-2-(3,5-Dimethoxyphenyl)-1-(4-hydroxyphenyl)ethene, (E)-4-(3,5-Dimethoxystyryl)phenol, (E)-4-Hydroxy-3',5'-dimethoxystilbene, 3,5-Dimethoxy-4'-hydroxy-trans-stilbene, 4-[(1E)-2-(3,5-Dimethoxyphenyl)ethenyl]phenol, E-Pterostilbene, Pterostilbene, trans-3,5-Dimethoxy-4'-hydroxystilbene, trans-Pterostilbene
IUPAC navn 4-[(E)-2-(3,5-dimethoxyphenyl)ethenyl]phenol
SMIL COc1cc(OC)cc(\C=C\c2ccc(O)cc2)c1
Tekniske data
Kemisk navn eller materiale Pterostilbene
Renhedsgrad noter HPLC
Mængde 1 g
InChI formel InChI=1S/C16H16O3/c1-18-15-9-13(10-16(11-15)19-2)4-3-12-5-7-14(17)8-6-12/h3-11,17H,1-2H3/b4-3+
Formel vægt 256.1099
Procent renhed >95%