Få mere at vide
SHP Steriltechnik AG PT100 Media Sensor
Artikelnummer. 31908807
Skift visning
Til brug med (udstyr):
Autoclaves
Artikelnummer. Til brug med (udstyr)
31908807 Autoclaves
1 options
Artikelnummer. 31908807 Leverandør SHP Steriltechnik AG Leverandørnr. 20.0000.000.004
 Begrænset vare
Forventet forsendelse: 30-11-2026
PT100 media sensor provides precise temperature measurement in liquids and gases, robust design for industrial applications, fast response time and high long-term stability.
  • Measuring range: typically -50° C to +250°C
  • Accuracy: Class A (±0.15° C at 0° C)
  • Fast response time thanks to compact sensor design
  • Robust stainless steel housing for a long service life
  • Connection options: cable or connector
  • Suitable for liquids, gases and aggressive media
Tekniske data
Til brug med (udstyr) Autoclaves
Model- Second Media sensor
Produkttype Media Sensor