Få mere at vide
Tocris Bioscience™ PT-65
Beskrivelse
PT-65 is a potent GSK3 Degrader (PROTAC®) (DC50 = 28.3 nM and 34.2 nM against GSK3α and GSK3β, respectively, in SH-SY5Y cells). It suppresses Okadaic acid (Cat. No. 1136) -induced tau hyperphosphorylation and ameliorates learning and memory impairments in rat models of Alzheimer's disease.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-Amino-6-(4-((4-(4-(1-(17-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)-3,6,9,12,15-pentaoxaheptadecyl)-1 H-1,2,3-triazol-4-yl)butyl)piperazin-1-yl)sulfonyl)phenyl)-N-(pyridin-3-yl)pyrazine-2-carboxamide |
| CAS | 2721998-87-2 |
| Mængde | 5 mg |
| Mål | Active Degraders |
| Molekylær formel | C51H63N13O12S |
| Renhed | 0.98 |