Learn More
BINDER™ Pt 100 Temperature Sensor
For Use With: BINDER units BD 56, BD 115, BD 260, BD 720
Brand: BINDER™ 8012-0940
This item is not returnable.
View return policy
Description
- Flexible
- Additional sensor
- Interior
Specifications
Pt 100 temperature sensor - additional flexible Pt 100, interior, for displaying the temperature on the unit display | |
BINDER Chambers BD 56, BD 115, BD 260, BD 720 | |
Plastic | |
Temperature Sensor |
Product Content Correction
Product Title
BINDER™ Pt 100 Temperature Sensor > For Use With: BD 56, BD 115, BD 260
Spot an opportunity for improvement?Share a Content Correction