Learn More
BINDER™ Pt 100 Temperature Sensor
For Use With: FD 56, FD 115, FD 260, FD 720
Brand: BINDER™ 8012-1620
This item is not returnable.
View return policy
Description
- Flexible
- Additional sensor
- Interior
Specifications
Pt 100 temperature sensor - additional flexible Pt 100, interior, for displaying the temperature on the unit display | |
BINDER Chambers FD 56, FD 115, FD 260, FD 720 | |
Plastic | |
Temperature Sensor |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BINDER™ Pt 100 Temperature Sensor > For Use With: FD 56, FD 115, FD 260, FD 720
Spot an opportunity for improvement?Share a Content Correction