missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
PSP (REG1A) Monoclonal Antibody (OTI13C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF813996
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
PSP (REG1A) Monoclonal antibody specifically detects PSP (REG1A) in Human samples. It is validated for ELISATekniske data
| PSP (REG1A) | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P05164 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4353 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| ELISA | |
| OTI13C9 | |
| Unconjugated | |
| MPO | |
| ICRF; islet cells regeneration factor; Islet of Langerhans regenerating protein; LITHOST; Lithostathine; lithostathine 1 alpha; lithostathine-1-alpha; MGC12447; P19; pancreatic stone protein; pancreatic stone protein, secretory; pancreatic thread protein; protein-X; PSP; PSPS; PSPS1; PTP; RATLITHOST; RATRGPI; Reg; Reg1; REG1A; REG-1-alpha; regenerating family member 1 alpha; regenerating islet-derived 1 alpha; regenerating islet-derived mouse homolog 1; regenerating islet-derived protein 1-alpha; Regenerating protein I alpha; regeneration protein, lithostatin, pancreatic stone protein; Rgp1; RGPI | |
| Full length recombinant protein of human MPO (NP_000241) produced in Baculovirus-Insect cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion