missing translation for 'onlineSavingsMsg'
Få mere at vide

Tocris Bioscience™ Psoralen-triethylene glycol azide
GREENER_CHOICE

Artikelnummer. 18739186
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18739186 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18739186 Leverandør Tocris Bioscience™ Leverandørnr. 6862/5

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

RNA conformation probe; reagent for COMRADES RNA platform

Psoralen TEG Azide is used to probe RNA structure and conformation in live cells, using the cross-linking of matched RNAs and deep sequencing or COMRADES method. Psoralen crosslinks the RNA and the azide enables subsequent click reaction with biotin. The COMRADES method has been used to determine the architecture of the Zika virus RNA genome inside cells, and identified multiple site-specific interactions with human noncoding RNAs. It has also been used to map RNA-RNA interactions along the SARS-CoV-2 genome and subgenomes inside cells. Psoralen TEG Azide is cell permeable. For detailed methods please see references section below.
TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 3-(13-Azido-5,8,11-trioxa-2-azatridec-1-yl)-2,5,9-trimethyl-7 H-furo[3,2-g][1 ]benzopyran-7-one
CAS 1352815-11-2
Mængde 5 mg
Mål DNA, RNA and Protein Synthesis Compounds
Molekylær formel C23H30N4O6
Renhed 0.98
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.