missing translation for 'onlineSavingsMsg'
Få mere at vide

PSN632408, MedChemExpress

Artikelnummer. 16469657
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16469657 5 mg
16467118 10 mg
16443157 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16469657 Leverandør Medchem Express Leverandørnr. HY16673

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress PSN632408, a selective, orally active GPR119 agonist, shows similar potency to OEA at both recombinant mouse and human GPR119 receptors (EC50=5.6 and 7.9 uM, respectively). PSN632408 can stimulate β-cell replication and improve islet graft function. PSN632408 has the potential for the research of obesity and related metabolic disorders.

PSN632408, a selective, orally active GPR119 agonist, shows similar potency to OEA at both recombinant mouse and human GPR119 receptors (EC50=5.6 and 7.9 uM, respectively). PSN632408 can stimulate β-cell replication and improve islet graft function. PSN632408 has the potential for the research of obesity and related metabolic disorders.

Tekniske data

Kemisk navn eller materiale PSN632408
Fysisk form Solid
CAS 857652-30-3
Farve White
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C18H24N4O4
Til brug med (applikation) Metabolism-sugar/lipid metabolism
Opløselighedsinformation DMSO : 50 mg/mL (138.73 mM; Need ultrasonic)
SMIL O=C(OC(C)(C)C)N1CCC(CC1)OCC2=NC(C3=CC=NC=C3)=NO2
Molekylvægt (g/mol) 360.41
Formel vægt 360.41
Procent renhed 99.64%
Grad Research
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.