missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ PSMD7 (Human) Recombinant Protein

Human PSMD7 full-length ORF ( AAH12606, 1 a.a. - 324 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00005713-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179821

  • 2735.00 DKK / 10µg
Estimated Shipment: 19-05-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MPELAVQKVVVHPLVLLSVVDHFNRIGKVGNQKRVVGVLLGSWQKKVLDVSNSFAVPFDEDDKDDSVWFLDHDYLENMYGMFKKVNARERIVGWYHTGPKLHKNDIAINELMKRYCPNSVLVIIDVKPKDLGLPTEAYISVEEVHDDGTPTSKTFEHVTSEIGAEEAEEVGVEHLLRDIKDTTVGTLSQRITNQVHGLKGLNSKLLDIRSYLEKVATGKLPINHQIIYQLQDVFNLLPDVSLQEFVKAFYLKTNDQMVVVYLASLIRSVVALHNLINNKIADRDAEKKEGQEKEESKKDRKEDKEKDKDKEKSDVKKEEKKEKK
Specifications

Specifications

AAH12606
proteasome (prosome, macropain) 26S subunit, non-ATPase, 7
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PSMD7
GST
5713
Wheat germ expression system
10 μg
MOV34, P40, Rpn8, S12
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers