Få mere at vide
Pseudooxynicotine Dihydrochloride (4-(Methylamino)-1-(3-pyridyl)butan-1-one Dihydrochloride), Mikromol
Artikelnummer. 30463998
Skift visning
Mængde:
25 mg
4 x 25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30463998 4 x 25 mg
30051359 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30463998 Leverandør Mikromol Leverandørnr. MM0517.1700254AMP
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Pseudooxynicotine Dihydrochloride (4-(Methylamino)-1-(3-pyridyl)butan-1-one Dihydrochloride), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-(Methylamino)-1-(3-pyridyl)-1-butanone Dihydrochloride
CAS 66093-90-1
Analyt- eller komponentnavne 4-(Methylamino)-1-(3-pyridyl)-1-butanone Dihydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C10 H14 N2 O . 2 Cl H
Certificeringer/compliance ISO 9001
Synonym Pseudooxynicotine Dihydrochloride, 1-Butanone, 4-(methylamino)-1-(3-pyridinyl)-, hydrochloride (1:2), 1-Butanone, 4-(methylamino)-1-(3-pyridinyl)-, dihydrochloride (9CI), 1-Butanone, 4-methylamino-1-(3-pyridyl)-, dihydrochloride
InChI formel InChI=1S/C10H14N2O.2ClH/c1-11-6-3-5-10(13)9-4-2-7-12-8-9;;/h2,4,7-8,11H,3,5-6H2,1H3;2*1H
Løsningstype Neat
SMIL Cl.Cl.CNCCCC(=O)c1cccnc1
IUPAC navn 4-(methylamino)-1-pyridin-3-ylbutan-1-one;dihydrochloride
Molekylvægt (g/mol) 251.15
Mængde 4 x 25 mg
Formel vægt 250.064 g/mol