Få mere at vide
Pseudoginsenoside F11, TRC
Artikelnummer. 17992895
Skift visning
Mængde:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17992895 100 mg
17944655 25 mg
17962995 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17992895 Leverandør Toronto Research Chemicals Leverandørnr. TRCP839550100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 69884-00-0
Molekylær formel C42 H72 O14
Molekylvægt (g/mol) 801.01
Synonym β-D-Glucopyranoside, (3β,6α,12β,24R)-20,24-epoxy-3,12,25-trihydroxydammaran-6-yl 2-O-(6-deoxy-α-L-mannopyranosyl)-, Dammarane, β-D-glucopyranoside deriv., (3β,6α,12β,24R)-20,24-Epoxy-3,12,25-trihydroxydammaran-6-yl 2-O-(6-deoxy-α-L-mannopyranosyl)-β-D-glucopyranoside, Ginsenoside A1, Pseudoginsenoside F11
IUPAC navn (2S,3R,4R,5R,6S)-2-[(2R,3R,4S,5S,6R)-2-[[(3S,5R,6S,8R,9R,10R,12R,13R,14R,17S)-3,12-dihydroxy-17-[(2S,5R)-5-(2-hydroxypropan-2-yl)-2-methyloxolan-2-yl]-4,4,8,10,14-pentamethyl-2,3,5,6,7,9,11,12,13,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-6-yl]oxy]-4,5-dihydroxy-6-(hydroxymethyl)oxan-3-yl]oxy-6-methyloxane-3,4,5-triol
SMIL C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2O[C@H]3C[C@]4(C)[C@H](C[C@@H](O)[C@@H]5[C@H](CC[C@@]45C)[C@]6(C)CC[C@@H](O6)C(C)(C)O)[C@@]7(C)CC[C@H](O)C(C)(C)[C@H]37)[C@H](O)[C@H](O)[C@H]1O
Tekniske data
Kemisk navn eller materiale Pseudoginsenoside F11
Mængde 100 mg
InChI formel InChI=1S/C42H72O14/c1-19-28(46)30(48)32(50)35(52-19)55-33-31(49)29(47)23(18-43)54-36(33)53-22-17-41(8)24(39(6)13-11-25(45)37(2,3)34(22)39)16-21(44)27-20(10-14-40(27,41)7)42(9)15-12-26(56-42)38(4,5)51/h19-36,43-51H,10-18H2,1-9H3/t19-,20-,21+,22-,23+,24+,25-,26+,27-,28-,29+,30+,31-,32+,33+,34-,35-,36+,39+,40+,41+,42-/m0/s1
Formel vægt 800.4922