Få mere at vide
Psammaplin A, TRC
Artikelnummer. 30422553
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30422553 5 mg
30417288 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30422553 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8401735MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 110659-91-1
Molekylær formel C22H24Br2N4O6S2
Molekylvægt (g/mol) 664.39
Synonym (αE,α'E)-N,N'-(Dithiodi-2,1-ethanediyl)bis[3-bromo-4-hydroxy-α-(hydroxyimino)benzenepropanamide], (E,E)-Psammaplin A, Bisprasin, Psammaplin A, UVI 5000, (E,E)-N,N'-(Dithiodi-2,1-ethanediyl)bis[3-bromo-4-hydroxy-α-(hydroxyimino)-benzenepropanamide]
IUPAC navn (2E,2'E)-N,N'-(disulfanediylbis(ethane-2,1-diyl))bis(3-(3-bromo-4-hydroxyphenyl)-2-(hydroxyimino)propanamide)
SMIL O=C(NCCSSCCNC(/C(CC1=CC=C(O)C(Br)=C1)=N/O)=O)/C(CC2=CC=C(O)C(Br)=C2)=N/O
Tekniske data
Kemisk navn eller materiale Psammaplin A
Mængde 5 mg
InChI formel InChI=1S/C22H24Br2N4O6S2/c23-15-9-13(1-3-19(15)29)11-17(27-33)21(31)25-5-7-35-36-8-6-26-22(32)18(28-34)12-14-2-4-20(30)16(24)10-14/h1-4,9-10,29-30,33-34H,5-8,11-12H2,(H,25,31)(H,26,32)/b27-17+,28-18+
Formel vægt 661.95