missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prucalopride succinate, MedChemExpress

Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
Description
Prucalopride succinate is a selective, high affinity 5-HT4 receptor agonist with pKi of 8.6/8.1 for 5-HT4a/4b.
Specifications
Specifications
Chemical Name or Material | Prucalopride succinate |
CAS | 179474-85-2 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C22H32ClN3O7 |
Quantity | 50 mg |
Synonym | R-108512 |
Solubility Information | DMSO : 50 mg/mL (102.89 mM; Need ultrasonic) |
SMILES | O=C(O)CCC(O)=O.O=C(C1=C(OCC2)C2=C(N)C(Cl)=C1)NC3CCN(CCCOC)CC3 |
Molecular Weight (g/mol) | 485.96 |
Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Prucalopride succinate, MedChemExpress
Spot an opportunity for improvement?Share a Content Correction