Få mere at vide
Protionamide, LoGiCal
Artikelnummer. 30463839
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30463839 10 mg
1 options
Artikelnummer. 30463839 Leverandør LoGiCal Leverandørnr. LGCFOR0844.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Protionamide
CAS 14222-60-7
Analyt- eller komponentnavne Protionamide
Forsendelsestilstand Room Temperature
Molekylær formel C9 H12 N2 S
Certificeringer/compliance ISO 9001
Synonym 2-Propylthio-isonicotinamide, 1321TH, 2-Propyl-4-pyridinecarbothioamide, 2-Propylisonicotinylthioamide, 2-Propylthioisonicotinamide, 9778 R. P., Ektebin, Prothionamide, Protion, Protionamid, Protionamide, Protionizina, RP 9778, TH 1321, Tebeform, Trevintix, Tuberex, Ethionamide Impurity C
InChI formel InChI=1S/C9H12N2S/c1-2-3-8-6-7(9(10)12)4-5-11-8/h4-6H,2-3H2,1H3,(H2,10,12)
Løsningstype Neat
SMIL CCCc1cc(ccn1)C(=S)N
IUPAC navn 2-propylpyridine-4-carbothioamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 180.27
Mængde 10 mg
Formel vægt 180.0721 g/mol