Få mere at vide
Prothipendyl Hydrochloride Monohydrate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Prothipendyl Hydrochloride Monohydrate |
| CAS | 70145-94-7 |
| Analyt- eller komponentnavne | Prothipendyl Hydrochloride Monohydrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H19 N3 S . Cl H . H2 O |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 10H-Pyrido[3,2-b][1,4]benzothiazine, 10-(3-dimethylaminopropyl)-, hydrochloride, hydrate (6CI), 10H-Pyrido[3,2-b][1,4]benzothiazine-10-propanamine, N,N-dimethyl-, monohydrochloride, monohydrate (9CI), 10H-Pyrido[3,2-b][1,4]benzothiazine-10-propanamine, N,N-dimethyl-, hydrochloride, hydrate (1:1:1), Prothipendyl Hydrochloride Monohydrate |
| InChI formel | InChI=1S/C16H19N3S.ClH.H2O/c1-18(2)11-6-12-19-13-7-3-4-8-14(13)20-15-9-5-10-17-16(15)19;;/h3-5,7-10H,6,11-12H2,1-2H3;1H;1H2 |
| Løsningstype | Neat |
| SMIL | O.Cl.CN(C)CCCN1c2ccccc2Sc3cccnc13 |
| Vis mere |