Få mere at vide
Prothipendyl-d6 Hydrochloride, TRC
Artikelnummer. 30423747
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30423747 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30423747 Leverandør Toronto Research Chemicals Leverandørnr. TRCP83885210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Prothipendyl-d6 Hydrochloride
CAS 1346604-67-8
Molekylær formel C16H13D6N3S • HCl
Mængde 10 mg
Synonym N,N-(Dimethyl-d6)-10H-pyrido[3,2-b][1,4]benzothiazine-10-propanamine Hydrochloride, AY 56031-d6, Azacon-d6, D 206-d6, Dominal-d6 Hydrochloride, Prothipendyl-d6 Monohydrochloride, Tolnate-d6
InChI formel InChI=1S/C16H19N3S.ClH/c1-18(2)11-6-12-19-13-7-3-4-8-14(13)20-15-9-5-10-17-16(15)19;/h3-5,7-10H,6,11-12H2,1-2H3;1H/i1D3,2D3;
SMIL [2H]C([2H])([2H])N(CCCN1C2=CC=CC=C2SC3=C1N=CC=C3)C([2H])([2H])[2H].Cl
IUPAC navn 3-pyrido[3,2-b][1,4]benzothiazin-10-yl-N,N-bis(trideuteriomethyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 291.443646
Formel vægt 327.1443