Learn More
Thermo Scientific™ Proteinase K, Tritirachium album limber
Commonly used in molecular biology to digest protein and remove contamination from preparations of nucleic acid. Addition of Proteinase K to nucleic acid preparations rapidly inactivates nucleases that might otherwise degrade the DNA or RNA during purification.
378.00 DKK - 1009.00 DKK
Specifications
Content And Storage Keep cold
Form Lyophilized Powder
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15474509
SDS View Documents Promotion Details
Thermo Scientific™
J62051.MB
25 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15484509
SDS View Documents Promotion Details
Thermo Scientific™
J62051.MC
100 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Keep cold
Lyophilized Powder