Få mere at vide
Molecular Probes™ Protein-Protein Crosslinking Kit
Artikelnummer. 10194752
Skift visning
Mængde:
1 Kit
Pakningsstørrelse:
1 sæt
Artikelnummer. Mængde unitSize
10194752 1 Kit 1 sæt
1 options
Artikelnummer. 10194752 Leverandør Molecular Probes™ Leverandørnr. P6305
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Protein-Protein Crosslinking Kit
The Protein-Protein Crosslinking Kit provides all of the reagents and purification media required to perform three protein-protein conjugations in which neither protein contains available thiol residues. Briefly, the amines of one of the proteins is thiolated with SPDP and the amines of the second protein are converted to thiol-reactive maleimides with SMCC. This kit was designed to prepare and purify protein-protein conjugates, including crosslinking phycobiliproteins (R-PE) to antibodies, but it can be modified to generate peptide-protein or enzyme-nucleic acid conjugates.
TRUSTED_SUSTAINABILITY
Tekniske data
Mængde 1 Kit