Learn More
Technical Service Consultants™ Protect™ Bacterial Preservation System, Gamma Irradiation Sterile
Use Technical Service Consultants™ Protect™ Bacterial Preservation System, Gamma Irradiation Sterile for long-term storage of a wide range of bacteria, yeasts and fungi. Vials for frozen storage are available in blue, white, green, yellow and red.
Brand: Technical Service Consultants™ TS/80-MX
Additional Details : Weight : 0.39240kg
Alert:
Initial cell concentration is directly related to satisfactory bead recovery; low inoculum can reduce vial longevity.
Description
- Multi-purpose cryobeads in mixed colors
- Micro-organism preservation Beads in Cryovial
- Mixed colored vials packed in moisture resistant box with user reference in box and on lid
- 80 vials in a box
Specifications
SAL 10-6, by Gamma-irradiation | |
Cryovials containing ceramic beads plus a cryopreservativefluid | |
For unopened vials: 4-25°C; in dry and ventilated area; protect from light, heat and moisture in tightly closed original container |
Mixed colors | |
Plastisized Paper Box |