Learn More
Protease-Free Bovine Albumin, Thermo Scientific Chemicals
Derived from bovine blood.
Brand: Thermo Scientific J10867.09
Additional Details : CAS Number : 9048-46-8 Weight : 1.00000kg
Description
- Purity grade: Molecular Biology Grade
- Assay (electrophoresis): ≥98.0%
- Protein: ≥96.0%
- MDLNumber: MFCD00130384
- CAS: 9048-46-8
Specifications
Albumin | |
10 g | |
4°C | |
Lyophilized |
6.8 to 7.2 | |
Bovine blood | |
Plastic bottle |
RUO – Research Use Only