missing translation for 'onlineSavingsMsg'
Learn More

Affymetrix/USB™ Protease-Free Bovine Albumin

Derived from bovine blood.

Brand:  Affymetrix/USB™ J10867.18

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15865068

  • 2765.62 DKK / 50g


Estimated Shipment date
from Supplier 18-03-2019
Add to basket

Description

Description

  • Purity grade: Molecular Biology Grade
  • Assay (electrophoresis): ≥98.0%
  • Protein: ≥96.0%
  • MDLNumber: MFCD00130384
  • CAS: 9048-46-8
Specifications

Specifications

Lyophilized powder
6.8 to 7.2
Bovine blood
Plastic bottle
Albumin
50g
4°C
Videos