Få mere at vide
Propyzamide-d3 (phenyl-2,4,6-d3), CDN
Artikelnummer. 30323534
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323534 0.005 g
30324588 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323534 Leverandør CDN Leverandørnr. CDND64310.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propyzamide D3 (phenyl-2,4,6 D3)
CAS 1219805-79-4
Molekylær formel C12 2H3 H8 Cl2 N O
Mængde 0.005 g
Synonym Propyzamide D3 (phenyl-2,4,6 D3), 3,5-Dichloro-N-(1,1-dimethyl-2-propyn-1-yl)-2,4,6-trideuteriobenzamide
InChI formel InChI=1 S/C12H11Cl2NO/c1-4-12(2,3)15-11(16)8-5-9(13)7-10(14)6-8/h1,5-7 H,2-3H3,(H,15,16)/i5D,6 D,7 D
SMIL [2 H]c1c(Cl)c([2 H])c(C(=O)NC(C)(C)C#C)c([2 H])c1Cl
IUPAC navn 3,5-dichloro-2,4,6-trideuterio-N-(2-methylbut-3-yn-2-yl)benzamide
Molekylvægt (g/mol) 259.15
Formel vægt 258.0406 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity