Få mere at vide
Propyrisulfuron, TRC
Artikelnummer. 30421387
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30421387 5 mg
30412973 50 mg
30418275 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30421387 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8393885MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 570415-88-2
Molekylær formel C16 H18 Cl N7 O5 S
Molekylvægt (g/mol) 455.88
Synonym 2-Chloro-N-[[(4,6-dimethoxy-2-pyrimidinyl)amino]carbonyl]-6-propylimidazo[1,2-b]pyridazine-3-sulfonamide, 1-[(2-Chloro-6-propylimidazo[1,2-b]pyridazin-3-yl)sulfonyl]-3-(4,6-dimethoxypyrimidin-2-yl)urea, Propyrisulfuron, TH 547, Zeta-One
IUPAC navn 1-(2-chloro-6-propylimidazo[1,2-b]pyridazin-3-yl)sulfonyl-3-(4,6-dimethoxypyrimidin-2-yl)urea
SMIL CCCc1ccc2nc(Cl)c(n2n1)S(=O)(=O)NC(=O)Nc3nc(OC)cc(OC)n3
Tekniske data
Kemisk navn eller materiale Propyrisulfuron
Mængde 5 mg
InChI formel InChI=1S/C16H18ClN7O5S/c1-4-5-9-6-7-10-18-13(17)14(24(10)22-9)30(26,27)23-16(25)21-15-19-11(28-2)8-12(20-15)29-3/h6-8H,4-5H2,1-3H3,(H2,19,20,21,23,25)
Formel vægt 455.0779