Learn More
Propylthiouracil, Mikromol™
Product Code. 17961166
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity Unit Size
17961166 250 mg 250mg
1 options
Product Code. 17961166 Supplier Mikromol Supplier No. MM3284.00
 Restricted Item
Forventet forsendelse: 30-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Propylthiouracil
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6-Propyl-2-thiouracil
CAS 51-52-5
Analyte or Component Names 6-Propyl-2-thiouracil
Shipping Condition Room Temperature
Molecular Formula C7 H10 N2 O S
Certifications/Compliance ISO 17034
Synonym Propylthiouracil, 2,3-dihydro-6-propyl-2-thioxopyrimidin-4(1H)-one
InChI Formula InChI=1S/C7H10N2OS/c1-2-3-5-4-6(10)9-7(11)8-5/h4H,2-3H2,1H3,(H2,8,9,10,11)
SMILES CCCC1=CC(=O)NC(=S)N1
IUPAC Name 6-propyl-2-sulfanylidene-1H-pyrimidin-4-one
Molecular Weight (g/mol) 170.23
Quantity 250 mg
Formula Weight 170.0514 g/mol