Få mere at vide
Propylparaben-d4, TRC
Artikelnummer. 30409279
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30409279 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30409279 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8382895MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxybenzoic-2,3,5,6-D4 Acid Propyl Ester
CAS 1219802-67-1
Renhedsgrad noter HPLC
Molekylær formel C10 H12 O3
Mængde 5 mg
Synonym 4-Hydroxybenzoic acid-propyl ester D4
InChI formel InChI=1S/C10H12O3/c1-2-7-13-10(12)8-3-5-9(11)6-4-8/h3-6,11H,2,7H2,1H3/i3D,4D,5D,6D
SMIL [2H]c1c([2H])c(C(=O)OCCC)c([2H])c([2H])c1O
IUPAC navn propyl 2,3,5,6-tetradeuterio-4-hydroxybenzoate
Molekylvægt (g/mol) 184.2251
Formel vægt 184.1038
Procent renhed >95