Beskrivelse
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Tekniske data
Tekniske data
| Emballage | Glass Bottle |
| CAS | 125306-83-4 |
| Fysisk form | Crystalline |
| Industry Type | Herbicides |
| Mængde | 50 mg |
| Grad | Analytical Standard |