Få mere at vide
Propoxyphenyl Thiosildenafil, TRC
Artikelnummer. 30409593
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30409593 5 mg
30428410 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30409593 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8316355MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 479073-87-5
Molekylær formel C23H32N6O3S2
Molekylvægt (g/mol) 504.67
Synonym 1,6-Dihydro-1-methyl-5-[5-[(4-methyl-1-piperazinyl)sulfonyl]-2-propoxyphenyl]-3-propyl-7H-pyrazolo[4,3-d]pyrimidine-7-thione, 1-[[3-(4,7-Dihydro-1-methyl-3-propyl-7-thioxo-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-propoxyphenyl]sulfonyl]-4-methyl-piperazine, rel-5-[5-[[(3R,5S)-3,5-Dimethyl-1-piperazinyl]sulfonyl]-2-propoxyphenyl]-1,6-dihydro-1-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidine-7-thione
SMIL S=C1C2=C(C(CCC)=NN2C)NC(C3=CC(S(N4CCN(C)CC4)(=O)=O)=CC=C3OCCC)=N1
Tekniske data
Kemisk navn eller materiale Propoxyphenyl Thiosildenafil
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C23H32N6O3S2/c1-5-7-18-20-21(28(4)26-18)23(33)25-22(24-20)17-15-16(8-9-19(17)32-14-6-2)34(30,31)29-12-10-27(3)11-13-29/h8-9,15H,5-7,10-14H2,1-4H3,(H,24,25,33)
Formel vægt 504.1977
Procent renhed >95%