Få mere at vide
Propoxycarbazone Sodium Salt, TRC
Artikelnummer. 30137891
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137891 250 mg
30135365 500 mg
17992695 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137891 Leverandør Toronto Research Chemicals Leverandørnr. TRCP831308250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 181274-15-7
Molekylær formel C15 H17 N4 O7 S . Na
Molekylvægt (g/mol) 420.37
Synonym Benzoic acid, 2-[[[(4,5-dihydro-4-methyl-5-oxo-3-propoxy-1H-1,2,4-triazol-1-yl)carbonyl]amino]sulfonyl]-, methyl ester, sodium salt (9CI), Attribut, BAY-MKH 6561, MKH 6561, Olympus, Procarbazone sodium, Propoxycarbazone sodium
IUPAC navn sodium;(2-methoxycarbonylphenyl)sulfonyl-(4-methyl-5-oxo-3-propoxy-1,2,4-triazole-1-carbonyl)azanide
SMIL [Na+].CCCOC1=NN(C(=O)[N-]S(=O)(=O)c2ccccc2C(=O)OC)C(=O)N1C
Tekniske data
Kemisk navn eller materiale Propoxycarbazone-sodium
Mængde 250 mg
InChI formel InChI=1S/C15H18N4O7S.Na/c1-4-9-26-14-16-19(15(22)18(14)2)13(21)17-27(23,24)11-8-6-5-7-10(11)12(20)25-3;/h5-8H,4,9H2,1-3H3,(H,17,21);/q;+1/p-1
Formel vægt 420.0716