Få mere at vide
Propoxur-d3 (N-Methyl-d3), TRC
Artikelnummer. 30409122
Skift visning
Mængde:
0.5 mg
Pakningsstørrelse:
0.5mg
Artikelnummer. Mængde unitSize
30409122 0.5 mg 0.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30409122 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8308060.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propoxur D3 (N-methyl D3)
CAS 1219798-56-7
Molekylær formel C11 2H3 H12 N O3
Mængde 0.5 mg
Synonym Propoxur D3 (N-methyl D3), Propoxur D3 (methyl D3)
InChI formel InChI=1S/C11H15NO3/c1-8(2)14-9-6-4-5-7-10(9)15-11(13)12-3/h4-8H,1-3H3,(H,12,13)/i3D3
SMIL [2H]C([2H])([2H])NC(=O)Oc1ccccc1OC(C)C
IUPAC navn (2-propan-2-yloxyphenyl) N-(trideuteriomethyl)carbamate
Molekylvægt (g/mol) 212.26
Formel vægt 212.124