Få mere at vide
Propofol-d17, TRC
Artikelnummer. 30428218
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30428218 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30428218 Leverandør Toronto Research Chemicals Leverandørnr. TRCP8297525MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propofol-D17
CAS 1261393-54-7
Renhedsgrad noter GC
Molekylær formel C12HD17O
Mængde 5 mg
Synonym Phen-3,4,5-d3-ol','2,6-bis[1-(methyl-d3)ethyl-1,2,2,2-d4]- (ACI), 2,6-Bis[1-(methyl-d3)ethyl-1,2,2,2-d4]phen-3,4,5-d3-ol (ACI), Propofol-d17, Propofol-D17, 2,6-Di-iso-propylphenol-d17
InChI formel InChI=1S/C12H18O/c1-8(2)10-6-5-7-11(9(3)4)12(10)13/h5-9,13H,1-4H3/i1D3,2D3,3D3,4D3,5D,6D,7D,8D,9D
SMIL [2H]c1c([2H])c(c(O)c(c1[2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
Molekylvægt (g/mol) 195.38
Formel vægt 195.2425
Procent renhed >95