Få mere at vide
Propofol beta-D-Glucuronide, TRC
Artikelnummer. 30136078
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136078 10 mg
30009460 1 mg
30007930 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136078 Leverandør Toronto Research Chemicals Leverandørnr. TRCP82978010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 114991-26-3
Molekylær formel C18 H26 O7
Molekylvægt (g/mol) 354.39
Synonym 2,6-Bis(1-methylethyl)phenyl beta-D-Glucopyranosiduronic Acid, Propofol Glucuronide;
IUPAC navn (2S,3S,4S,5R,6S)-6-[2,6-di(propan-2-yl)phenoxy]-3,4,5-trihydroxyoxane-2-carboxylic acid
SMIL CC(C)c1cccc(C(C)C)c1O[C@@H]2O[C@@H]([C@@H](O)[C@H](O)[C@H]2O)C(=O)O
Tekniske data
Kemisk navn eller materiale Propofol Glucuronide
Renhedsgrad noter HPLC
Mængde 10 mg
InChI formel InChI=1S/C18H26O7/c1-8(2)10-6-5-7-11(9(3)4)15(10)24-18-14(21)12(19)13(20)16(25-18)17(22)23/h5-9,12-14,16,18-21H,1-4H3,(H,22,23)/t12-,13-,14+,16-,18+/m0/s1
Formel vægt 354.1679
Procent renhed >95%