Få mere at vide
Propiverine Hydrochloride, Mikromol™
Beskrivelse
Propiverine Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Propiverine, Hydrochloride |
| CAS | 54556-98-8 |
| Analyt- eller komponentnavne | Propiverine, Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C23 H29 N O3 . Cl H |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Acetic acid, diphenylpropoxy-, 1-methyl-4-piperidyl ester, hydrochloride (7CI), Mictonetten, Mictonorm, P 4, P 4 (anticholinergic), Pollarine, Propiverine hydrochloride, alpha-phenyl-alpha-propoxy-Benzeneacetic acid 1-methyl-4-piperidinyl ester hydrochloride (1:1), Benzeneacetic acid, alpha-phenyl-alpha-propoxy-, 1-methyl-4-piperidinyl ester, hydrochloride (9CI) |
| InChI formel | InChI=1S/C23H29NO3.ClH/c1-3-18-26-23(19-10-6-4-7-11-19,20-12-8-5-9-13-20)22(25)27-21-14-16-24(2)17-15-21;/h4-13,21H,3,14-18H2,1-2H3;1H |
| SMIL | Cl.CCCOC(C(=O)OC1CCN(C)CC1)(c2ccccc2)c3ccccc3 |
| IUPAC navn | (1-methylpiperidin-4-yl) 2,2-diphenyl-2-propoxyacetate;hydrochloride |
| Vis mere |