Få mere at vide
Propisochlor, TRC
Artikelnummer. 30134834
Skift visning
Mængde:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134834 50 mg
17908719 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30134834 Leverandør Toronto Research Chemicals Leverandørnr. TRCP82803350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 86763-47-5
Molekylær formel C15 H22 Cl N O2
Molekylvægt (g/mol) 283.79
Synonym 2-Chloro-N-(2-ethyl-6-methylphenyl)-N-[(1-methylethoxy)methyl]acetamide, (±)-Propisochlor, Propisochlor, Propizochlor, Proponit, Proponit 720EC, Proponit 720SC
IUPAC navn 2-chloro-N-(2-ethyl-6-methylphenyl)-N-(propan-2-yloxymethyl)acetamide
SMIL CCc1cccc(C)c1N(COC(C)C)C(=O)CCl
Tekniske data
Kemisk navn eller materiale Propisochlor
Mængde 50 mg
InChI formel InChI=1S/C15H22ClNO2/c1-5-13-8-6-7-12(4)15(13)17(14(18)9-16)10-19-11(2)3/h6-8,11H,5,9-10H2,1-4H3
Formel vægt 283.1339