Få mere at vide
Propiophenone-d10, CDN
Artikelnummer. 30309849
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30309849 1 g
30325679 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309849 Leverandør CDN Leverandørnr. CDND58621G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propiophenone-D10
CAS 288400-96-4
Molekylær formel C9 D10 O
Mængde 1 g
Synonym 1-Propanone-2,2,3,3,3-d5, 1-(phenyl-d5)- (9 CI), Propiophenone-d10, Propiophenone-D10, 1-(Phenyl-d5)-1-propanone-2,2,3,3,3-d5
InChI formel InChI=1 S/C9H10O/c1-2-9(10)8-6-4-3-5-7-8/h3-7 H,2H2,1H3/i1D3,2D2,3 D,4 D,5 D,6 D,7 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)C([2 H])([2 H])C([2 H])([2 H])[2 H])c([2 H])c1[2 H]
IUPAC navn 2,2,3,3,3-pentadeuterio-1-(2,3,4,5,6-pentadeuteriophenyl)propan-1-one
Molekylvægt (g/mol) 144.237
Formel vægt 144.136 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity