Få mere at vide
Propiophenone-2',3',4',5',6'-d5, CDN
Artikelnummer. 30309484
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30309484 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309484 Leverandør CDN Leverandørnr. CDND18151G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propiophenone-d5
CAS 54419-23-7
Molekylær formel C9 D5 H5 O
Mængde 1 g
Synonym 1-Propanone, 1-(phenyl-d5)- (9 CI), Propiophenone-d5, Propiophenone-2',3',4',5',6'-d5, Propiophenone-D5 (phenyl-D5), 1-(Phenyl-d5)-1-propanone
InChI formel InChI=1 S/C9H10O/c1-2-9(10)8-6-4-3-5-7-8/h3-7 H,2H2,1H3/i3D,4 D,5 D,6 D,7 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)CC)c([2 H])c1[2 H]
IUPAC navn 1-(2,3,4,5,6-pentadeuteriophenyl)propan-1-one
Molekylvægt (g/mol) 139.206
Formel vægt 139.105 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity