Få mere at vide
Propionylpromazine-d6 Hydrochloride, TRC
Artikelnummer. 30425085
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30425085 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425085 Leverandør Toronto Research Chemicals Leverandørnr. TRCP82750210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propionylpromazine D6 (N-dimethyl D6) Hydrochloride
CAS 1262770-67-1
Molekylær formel C20 H18 D6 N2 OS . HCl
Mængde 10 mg
Synonym 1-[10-[3-(Dimethylamino-d6)propyl]-10H-phenothiazin-2-yl]-1-propanone Hydrochloride, 10-[3-(Dimethyl-d6)aminopropyl]-2-propionylphenothiazine Hydrochloride, 1497CB-d6, Combelen-d6, Combilen-d6, 2-Propionyl-10-[3- (dimethyl-d6)aminopropyl)phenothiazine Hydrochloride, Propionpromazine-d6 Hydrochloride, Propiopromazine-d6 Hydrochloride, Tranvet-d6
InChI formel InChI=1S/C20H24N2OS.ClH/c1-4-18(23)15-10-11-20-17(14-15)22(13-7-12-21(2)3)16-8-5-6-9-19(16)24-20;/h5-6,8-11,14H,4,7,12-13H2,1-3H3;1H/i2D3,3D3;
SMIL C(CCN(C([2H])([2H])[2H])C([2H])([2H])[2H])N1C=2C(SC=3C1=CC=CC3)=CC=C(C(CC)=O)C2.Cl
IUPAC navn 1-[10-[3-[bis(trideuteriomethyl)amino]propyl]phenothiazin-2-yl]propan-1-one;hydrochloride
Molekylvægt (g/mol) 346.523646
Formel vægt 382.1753