Få mere at vide
Propionic-d2 Acid, TRC
Artikelnummer. 30412210
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30412210 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412210 Leverandør Toronto Research Chemicals Leverandørnr. TRCP786390500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propionic Acid 2,2-D2
CAS 19136-91-5
Molekylær formel C3 D2 H4 O2
Mængde 500 mg
Synonym Propanoic-2,2-d2 acid (9CI), Propionic-2,2-d2 acid (7CI','8CI), α-Dideuteropropanolic acid, Propionic acid D2 (methylene-D2), Propionic acid 2,2-D2
InChI formel InChI=1S/C3H6O2/c1-2-3(4)5/h2H2,1H3,(H,4,5)/i2D2
SMIL [2H]C([2H])(C)C(=O)O
IUPAC navn 2,2-dideuteriopropanoic acid
Molekylvægt (g/mol) 76.091
Formel vægt 76.049