Få mere at vide
Propionic Acid Methyl-d3 Ester, TRC
Artikelnummer. 30425646
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30425646 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30425646 Leverandør Toronto Research Chemicals Leverandørnr. TRCP786372100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propionic Acid Methyl-D3 Ester
CAS 38758-64-4
Molekylær formel C4 D3 H5 O2
Mængde 100 mg
Synonym Methyl Tetracosanoate-D3, Methan-d3-ol','propanoate (9CI), Propionic acid','methyl-d3 ester (6CI), Methyl-d3 propionate, Propionic acid methyl-d3 ester, Propanoic acid methyl-d3 ester, Propionic acid methyl-D3 ester
InChI formel InChI=1S/C4H8O2/c1-3-4(5)6-2/h3H2,1-2H3/i2D3
SMIL [2H]C([2H])([2H])OC(=O)CC
IUPAC navn trideuteriomethyl propanoate
Molekylvægt (g/mol) 91.124
Formel vægt 91.071