Få mere at vide
Propionic Acid-d3, TRC
Artikelnummer. 30414721
Skift visning
Mængde:
2.5 g
Artikelnummer. Mængde
30414721 2.5 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414721 Leverandør Toronto Research Chemicals Leverandørnr. TRCP7863622.5G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Propionic Acid-d3
CAS 55577-88-3
Molekylær formel C3 D3 H3 O2
Mængde 2.5 g
Synonym Propanoic-3,3,3-d3 acid (9CI), Propionic-3,3,3-d3 acid (7CI), Propionic acid D3 (methyl D3), Propionic acid-d3, Propionic acid 3,3,3-D2
InChI formel InChI=1S/C3H6O2/c1-2-3(4)5/h2H2,1H3,(H,4,5)/i1D3
SMIL [2H]C([2H])([2H])CC(=O)O
IUPAC navn 3,3,3-trideuteriopropanoic acid
Molekylvægt (g/mol) 77.097
Formel vægt 77.056