Learn More
Propionaldehyde (>80%), TRC
Product Code. 30410875
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30410875 10 g -
30427124 5 g 5g
30415466 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30410875 Supplier Toronto Research Chemicals Supplier No. TRCP78251510G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 123-38-6
Molecular Formula C3 H6 O
Molecular Weight (g/mol) 58.08
Synonym Propanal, Propionaldehyde (7CI,8CI), 1-Propanal, Ethylcarboxaldehyde, Methylacetaldehyde, NSC 6493, Propaldehyde, Propanaldehyde, Propional, Propionic aldehyde, Propylaldehyde, Propylic aldehyde, n-Propanal, n-Propionaldehyde
IUPAC Name propanal
SMILES CCC=O
Specifications
Quantity 10 g
InChI Formula InChI=1S/C3H6O/c1-2-3-4/h3H,2H2,1H3
Formula Weight 58.0419
Chemical Name or Material Propionaldehyde